Learn More
Fisherbrand™ Foamed PE-Lined Polypropylene White Caps
For general purpose use. Fisherbrand™ Foamed PE-Lined Polypropylene White Caps offer good impact strength and pliability.
 View more versions of this product
Catalog No. 02-912-092
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • Liner molds itself around the sealing areas virtually eliminating leakage
  • Chemical resistant including acids, alcohols, alkalis, aqueous products, cosmetics, household oils and solvents
Description & Specifications
Specifications
ISO 9001
White
3500 Cs.
24-410
Foam Polyethylene
Plastic
Bottle