Thermo Scientific™ Fluorescent Antibody Microscope Slides Available on GSA/VA Contract for Federal Government customers only.

Allow for easy handling with Thermo Scientific™ Fluorescent Antibody Slides.

 View more versions of this product

Catalog No. 22-339408

  Check Availability
Add to Cart

    Specifications

    Packaging Quantity 72 slides per box, 20 boxes per case
    Description Light white Superfrost tab slide with two 10mm diameter circles
    Width (Metric) 25mm
    Corner Style 90°
    Dimensions (L x W) 75 x 25mm
    Thickness (Metric) 1mm
    Color Light white
    Edges Ground
    Length (Metric) 75mm

    Allow for easy handling with Thermo Scientific™ Fluorescent Antibody Microscope Slides. These slides feature a durable 3/4" light white Superfrost tab with two light white Superfrost circles. The printed tab and circles are impervious to virtually all common laboratory chemicals and reagents. Manufacturer Part Number 3032 is a premium Thermo Scientific™ Gold Seal™ Slide which uses a brilliant white epoxy ink for the slide tab and the two circles.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit