Learn More
Fisherbrand™ Full- and Mid-Size Faceshields
High-quality, optically clear, distortion-free 7mil film
 View more versions of this product
Catalog No. 19-460-101
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Polyester lens is treated with antifog and antistatic coating
  • Headpiece is held in place by a stretch strap and includes a latex-free foam band for comfortable extended wear
Compliance
Conform to OSHA 29CFR Part 1910.1030 for bloodborne pathogens and the CDC's recommendation for protective eyewear.
Specifications
Specifications
Conforms to OSHA 29CFR Part 1910.1030 for bloodborne pathogens and the CDC's recommendation for protective eyewear.
Polyester
Clear
Anti-fog, Anti-static
Mid-Faceshield
Anti-fog, Anti-static