Learn More
Fisherbrand Gas Cylinder Tags
Ideal for avoiding mishandling of cylinders
$20.95
Catalog Number | For Use With (Application) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | For Use With (Application) | Price | Quantity & Availability | ||||||
18-999-792
![]() ![]() |
For avoiding mishandling of cylinders |
Case of 100 for $20.95
|
![]() |
Description
Cylinder Status Tags- Simply tear off perforated sections to indicate if cylinder is full, in service, or empty
- Tags come with wire ties
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Wire ties |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.