Fisherbrand™ Gel-Loading Tips, 1-200µL - PROMO

An ideal choice for accurately and easily loading wells in gel electrophoresis

 View more versions of this product

Catalog No. 02-707-138PM

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Product Type 10 × 96
    Material Polypropylene
    Length (Metric) 6.98cm
    Volume (Metric) 1 to 200μL
    Sterility Nonsterile
    Tip Style Gel, Round
    Certifications/Compliance RNase, DNase-free
    For Use With (Application) For DNA sequencing or loading electrophoresis gels
    Color Natural

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit