Learn More
Fisherbrand? Gel-Loading Tips, 1-200µL - PROMO
An ideal choice for accurately and easily loading wells in gel electrophoresis
 View more versions of this product
Catalog No. 02-707-139PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
10 × 96
Sterile
Gel, Round
6.98cm
For DNA sequencing or loading electrophoresis gels
1 to 200μL
Natural
Polypropylene
RNase, DNase-free