Learn More
Fisherbrand™ General Filtration Membrane Filters Available on GSA/VA Contract for Federal Government customers only.
Useful for most filter applications
 View more versions of this product
Catalog No. 09-719-2E
Add to cart
Description
Description
  • Plain, white membrane filters
  • Pure, biologically inert mixture of cellulose acetate and cellulose nitrate (MCE)
  • Compatible with dilute acids and bases, aliphatic and aromatic hydrocarbons, and nonpolar liquids
  • Filters wet readily with water and other fluids
  • Contain no fibers or particles to contaminate filtrate
  • Autoclavable
Applications:
For all analytical, ultracleaning, and sterilizing applications at temperatures below 75°C (167°F)—except those involving esters, ether-alcohols, ketones, nitroparaffins or strong acids and bases
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
0.45μm
47mm
For all analytical, ultracleaning, and sterilizing applications at temperatures below 75°C (167°F) – except those involving esters, ether-alcohols, ketones, nitroparaffins or strong acids and bases.
Mixed Cellulose Esters (MCE) (Nitrate and Acetate)
Yes