Learn More
Fisherbrand™ General Purpose Liquid-in-Glass Partial-Immersion Thermometers - Promo
Special Offer: Save up to 50%
 View more versions of this product
Catalog No. 13-201-521PMO
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
2°C
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries
Partial
305mm
1°C
-100° to +50°C
ISO/IEC 17025:2005, A2LA, NIST
76mm
12 in.
Glass
Not Applicable
Blue Organic