Learn More
Fisherbrand™ General-Purpose Mercury Laboratory Thermometers: Partial-Immersion
Available in a variety of types for a wide range of applications
 View more versions of this product
Catalog No. 14-995-5G
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
These products contain mercury. Do not put products containing mercury in the trash. They should be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Specifications Description & Specifications
Specifications
76mm
405mm
20° to 750°F
15.9 in.
2°C
-10° to +400°C