Learn More
Fisherbrand™ General-Purpose Mercury Laboratory Thermometers: Total-Immersion
 View more versions of this product
Catalog No. 14-986B
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
These products contain mercury. Do not put products containing mercury in the trash. They should be recycled or disposed of as hazardous waste.
Specifications Description & Specifications
Specifications
Total
305mm
-20° to +110°C
12 in.
1°C