Learn More
Fisherbrand™ General-Purpose Mercury Laboratory Thermometers: Total-Immersion
Alert:
These products contain mercury. Do not put products containing mercury in the trash. They should be recycled or disposed of as hazardous waste.
Description
- With yellow and white back
Specifications
Total | |
405 mm | |
-10°C | |
1°C |
16 in. | |
260°C | |
-10°C to +260°C |