Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ General-Purpose Mercury Laboratory Thermometers: Total-Immersion
Specifications
Immersion Type | Total |
---|---|
Length (Metric) Overall | 405 mm |
Max. Temperature (Metric) | 260°C |
Min. Temperature (Metric) | -10°C |
Temperature Range (Metric) | -10°C to +260°C |
Alert:
These products contain mercury. Do not put products containing mercury in the trash. They should be recycled or disposed of as hazardous waste.
Catalog Number | Description | Price | Qty | Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Description | Price | Qty | Availability | |||||
14-983-10E
|
-10° to +260°C; Yellow back | -10° to +260°C; Yellow back | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
14-986E
|
-10° to +260°C; White back | -10° to +260°C; White back | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- With yellow and white back
Specifications
Total | |
260°C | |
-10°C to +260°C |
405 mm | |
-10°C | |
1°C |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title