Learn More
Fisherbrand™ General-Purpose Rubber Tubing in 12ft. (3.7m) Lengths
Seamless rubber tubing with nonslip surface and flexibility
Description
- Made with dry natural rubber
- Available in variety of sizes
- Offered in Black and Red
Specifications
3/8 in. | |
Rubber | |
14.2 mm | |
3.7 m | |
2.36 mm | |
Red |
9.65 mm | |
0.56 in. | |
12 ft. | |
0.093 in. | |
General purpose |