Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ GHS Labeled Right-to-Know Safety-Vented Wash Bottles

Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.

 View more versions of this product

Catalog No. 11-865-159



Add to cart

Description

Description

Unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips and allows dispensing of wash bottle in upright or inverted position.

  • Pre-printed bottles with chemical name and formula, GHS pictogram and signal word; National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Translucent LDPE bottles, Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm
  • Wide mouth 53mm (2 in.) for easy refilling
  • Color-coded polypropylene closures aid in quick and easy identification
  • Assortment bag includes one each: Acetone, Isopropanol, Bleach and Ethanol
Bottle with Label: TOLUENE
  • Red LDPE bottle
  • Does not include vented valve


Bottle with Label: DEIONIZED H2O, DISTILLED H20 and WATER
  • Does not indicate hazardous chemical and does not have/require GHS pictogram or signal word


Bottle with Label: METHYL ETHYL KETONE
  • Does not include vented valve

Compliance

Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA’s HazCom 2012 requirements – as pertains to “Workplace Labels”.

Specifications

Specifications

500mL
ISOTONIC SALINE
Natural
3/Bag
Natural
Wide mouth
500 mL
Polypropylene
LDPE, LDPE
No
53mm
Cap
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit