Learn More
Fisherbrand™ 51 Expansion Glass Shell Vials with Plug Style Closures Available on GSA/VA Contract for Federal Government customers only.
Straight-sided shell vials are ideal for dry contents.
 View more versions of this product
Catalog No. 03-339-26D
  Check Availability
Add to cart
Description
Description
Straight-sided shell vials are ideal for dry contents.
  • TITESEAL™ vials
  • Plain tops
  • Open-bottom plug style plastic closures, unattached
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Non-sterile
19mm
Yes
Borosilicate Glass
Shell Vial
Plain
65mm
3 dr.
Titeseal™
Clear
3 dr.
No
65mm
11.1mL
Ideal for storing dry products
Documents
Promotions
Promotions
  • Fisherbrand , Fisher Scientific , DWK Life Sciences (Kimble)
    Get up to $250 in glassware at no additional cost with your qualifying Fisherbrand™ Glassware purchase.
    06-30-2019