Learn More
Fisherbrand™ Graduated Valumetric™ Bottles with PTFE, Polyvinyl-Lined Caps
Store a variety of liquids and solids—dry powders, specimens and soil samples
 View more versions of this product
Catalog No. 02-911-453
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Clear, USP Type III soda-lime glass for easy viewing of contents
  • Printed graduations show milliliters and fluid ounces
  • Wide mouth for easy filling
Specifications
Specifications
1 oz., 30mL
1 oz.
Poly/Vinyl
Glass
48/Cs.
Screw
30mL
PTFE
Clear
48/Cs.
33-400