Learn More
Fisherbrand™ Graduated Valumetric™ Bottles without Caps
Store a variety of liquids and solids—dry powders, specimens and soil samples
 View more versions of this product
Catalog No. 02-912-119
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Clear, USP Type III soda-lime glass
  • Highly visible, white screened graduations in millimeters and ounces for general mixing and measuring applications
  • Wide mouth for easy entry and retrieval during storage and packaging applications
Specifications
Specifications
16 oz., 500mL
16 oz.
Soda Lime Glass
No
Wide
500mL
Clear
48/PK.
70-400
Screw