Learn More
Fisherbrand™ Hand Tally Counters with Six Counting Units
With six counting units
Catalog No. 22-363-567
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Five counting keys with 0 to 999 range
  • Totalizing counter records total number of strokes made on other keys; rings bell at increments of 100
  • Feature dual reset knobs