Fisherbrand™ Hanging Drop Slides
For examining living microorganisms such as bacteria and yeast in hanging drops
 View more versions of this product
Catalog No. 12-565A
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For examining living microorganisms such as bacteria and yeast in hanging drops
    No. of Wells 1
    Thickness 3.1 to 3.22mm
    Product Type Hanging Drop Slide
  • Description & Specifications
Related Products
Product Content Feedback