Fisherbrand™ HDPE Cylinder Rounds, With Polyethylene Foam-Lined Closures
Ideal for storing, shipping, and packaging liquids for long periods
 View more versions of this product
Catalog No. 02-912-294
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Capacity (Metric) 1000mL
    Capacity (English) 32 oz.
    Certifications/Compliance ISO 9001
    Closure Screw
    Closure Material Polyethylene
    Closure Liner Foam Polyethylene
    Closure Size 28-400
    Color Natural Translucent
    Material HDPE
    Neck Finish 28-400
    Capacity 32 oz., 1000mL (77/Cs.)
  • Description & Specifications
Related Products
Product Content Feedback