Fisherbrand™ HDPE Wide-Mouth Round Bottles
For storing solids or liquids
 View all options for this product
Catalog No. 02-912-104
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Capacity 1 oz., 30mL (1650/Pk.)
    Capacity (Metric) 30mL
    Capacity (English) 1 oz.
    Certifications/Compliance ISO 9001
    Closure Size 28-400
    Closure Type Screw
    Color Translucent
    Material HDPE
    Neck Finish 28-400
    • Translucent HDPE
    • Wide mouth for easy filling
  • Description & Specifications
Related Products
Product Content Feedback