Fisherbrand™ Histosette II™ Tissue and Biopsy Cassettes
Open and close as often as necessary — cassettes will always relock securely
 View more versions of this product
Catalog No. 15-182-709E
  Check Availability
Add to Cart
    Specifications
    Pore Style Slots
    Material Plastic
    Description Biopsy Cassette
    Writing Face Angle 45°
    Closure Type Separate Lid
    Color Blue
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback