Learn More
Fisherbrand™ Histosette™ II Tissue Processing/Embedding Cassettes with Separate Lid and Base
Histosette™ II Tissue Processing/Embedding Cassettes with Separate Lid and Base open and close as often as necessary - cassettes will always relock securely.
 View more versions of this product
Catalog No. 15-182-708C
Add to Cart
Description
Description
  • Disposable, plastic cassettes hold tissue specimens during embedding, processing, and storage
  • Molded from a special high-density polymer which is resistant to the chemical action of histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • The anterior writing surface is slanted at a 45° angle, making the cassette more suitable for use with certain types of labeling instruments
Order Info
Available in 11 noncytotoxic, nonmetallic colors.
Specifications
Specifications
Slots
Separate Lid
Plastic
45°
Green
Tissue Cassette
SDS
Documents
Documents
Product Certifications
Accessories