Fisherbrand™ HMIG and NFR Dispenser Labels Available on GSA/VA Contract for Federal Government customers only.

Keep workers safe by properly informing them of workplace hazards

 View more versions of this product

Catalog No. 18-999-785

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Keep workers safe by properly informing them of workplace hazards
    Product Type HMIG Dispenser Label
    Quantity 750 Pack
    Size (English) 0.87 x 0.87 in.
    Size (Metric) 0.02 x 02cm
    Format Label
    Color Assorted

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit