Learn More
Fisherbrand HMIG and NFR Write-On Labels, Hazardous Materials Identification Guide (HMIG) write on labels
Ideal for adding vital hazard information to surfaces of small bottles, cans, and other containers.
$64.90
Specifications
Color | Assorted |
---|---|
Format | Label |
Quantity | 1000/Roll |
For Use With (Application) | Ideal for adding vital hazard information to surfaces of small bottles, cans and other containers |
Length (Metric) | 9.8 cm |
Catalog Number | Product Type | Length (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Product Type | Length (English) | Price | Quantity & Availability | |||||
18-999-789
![]() |
NFR Write-On™ Label | 3.87 in. |
Case of 1000 for $64.90
|
|
18-999-788
![]() |
HMIG Write-On™ Label | 3.88 in. |
Case of 1000 for $64.90
|
|
Description
Ideal for adding vital hazard information to surfaces of small bottles, cans, and other containers.
- HMIG and NFR paper labels are large enough to include the chemical name, common name and manufacturer's name
- NFR labels let you write in the chemical name, owner, date, and hazard rating
Order Info
Both labels measure 3.87L x 1.50 in. W (9.8 x 3.8cm).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Assorted | |
1000/Roll | |
9.8 cm |
Label | |
Ideal for adding vital hazard information to surfaces of small bottles, cans and other containers | |
1.5 in. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.