Learn More
Fisherbrand™ Holders for Biohazard Autoclave Bags
Holds Fisherbrand™ and most other biohazard autoclave bags.
 View more versions of this product
Catalog No. 14-375-175
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Holders are not autoclavable.
Description
Description
  • High-density polyethylene container with rigid low-density polyethylene lid with knob
  • Lid fits securely to seal against hazardous aerosols
  • Containers have integral handle for easy transporting
  • For use on benchtop or floor
  • Accept bags with minimum width of 12, 16 and 20 in. (30.5, 40.6 and 50.8cm)
Specifications
Specifications
No
7.6L
HDPE (High-density Polyethylene)
28cm
8 qt.
Natural
11 in.
Lid
Documents
Accessories