Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Holders for Biohazard Autoclave Bags

Holds Fisherbrand™ and most other biohazard autoclave bags.

 View more versions of this product

Catalog No. 14-375-176



Add to cart

Alert:

Holders are not autoclavable.


Description

Description

  • High-density polyethylene container with rigid low-density polyethylene lid with knob
  • Lid fits securely to seal against hazardous aerosols
  • Containers have integral handle for easy transporting
  • For use on benchtop or floor
  • Accept bags with minimum width of 12, 16 and 20 in. (30.5, 40.6 and 50.8cm)

Specifications

Specifications

16 in.
Lid
24 qt.
Natural
41cm
No
22.7L
HDPE (High-density Polyethylene)
Documents
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit