Fisherbrand™ Holders for Biohazard Autoclave Bags

Hold Fisherbrand and most biohazard autoclave bags

 View more versions of this product

Catalog No. 14-375-177

  Check Availability
Add to Cart

Alert

Holders are not autoclavable.


    Specifications

    Height (Metric) 51cm
    Includes Lid
    Capacity (English) 48 qt.
    Capacity 48 qt., 50.8L
    Height (English) 20 in.
    Capacity (Metric) 50.8L
    Material HDPE

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit