Learn More
Fisherbrand™ Hollow Cathode Multi-Element 1 1/2in. Lamps
Window materials are quartz, borosilicate and UV glass; Lamps are multi-element and 1 1/2 in. (37mm) in diameter
 View more versions of this product
Catalog No. 14-386-103S
Add to cart
Description
Description
  • Quartz windows with neon gas fill (sodium/potassium has UV glass)
  • Lamps are multi-element and have 1 1/2 in. diameter (37mm) bodies For use in all AA spectrophotometers except Perkin-Elmer™ instruments
  • May be adapted to fit PE™ instruments using the 14-386-102Z Adapter Kit (sold separately)
Specifications
Specifications
2 Pin
3.7cm
UV Glass
Potassium, Sodium
Hollow Cathode
1.5 in.
Spectrophotometer
X Shape
Neon
Spectrophotometer Lamp