Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Fisherbrand™ IB™ Model Serum Filters
Store serum up to seven days when it is essential to prevent any exchange of ions.
 View more versions of this product
Catalog No. 02-681-67
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • One-way valve permits a physical gap to be created between the serum/plasma and clot, and prevents backflow into the primary collection tube
  • Use with glass or plastic collection tubes
  • Technique reduces second centrifugation, tube labeling and sample preparation time
  • Allows direct sampling into automatic laboratory equipment
  • Eliminates fibrin, particulate matter and equipment downtime
Specifications
Specifications
0.5 in.
Eliminates fibrin, particulate matter and equipment downtime
6 in.
100 Pack
Serum Filter
13mm
Glass or Plastic collection tubes
152mm
Polyethylene
Documents
Accessories