Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ IB™ Serum Filter
Allows for direct instrument sampling.
Description
- For use with 13mm plastic evacuated blood collection tubes
- Store plasma up to four days, serum up to seven days when it is essential to prevent any exchange of ions
- One-way valve permits leak-resistant seal between the clot and the filtered serum
- Life of gel tubes extended up to seven days without transfer to secondary tubes
- Filter removes fibrin and gel particulates
Specifications
100 Pack | |
0.5 in. | |
Polyethylene | |
Serum Filter | |
Removes fibrin and gel particulates |
13mm | |
1.62 in. | |
13mm Plastic evacuated Blood collection tubes | |
41mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction