Fisherbrand™ Industrial-Grade Vinyl Cleanroom Tape

For light industrial use, clean construction and cleanroom support

 View more versions of this product

Catalog No. 19-120-3210

  Check Availability
Add to Cart

    Specifications

    Format Tape
    Length (Metric) 33m
    Material Vinyl
    Product Type Industrial grade cleanroom Tape
    Quantity Each
    Color Brown
    Certifications/Compliance FDA, CID
    Width (English) 0.5 in.
    Grade Industrial
    For Use With (Application) Aisle marking, labeling, batching, clean-build projects, light industrial use and food processing applications
    Length (English) 36 yd.
    Sterile No

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit