Learn More
Fisherbrand™ Infant Heel Warmers
An easy, safe and reliable way to obtain better heelsticks
Specifications
Actuation Flex-disk
Product Type Infant Heel Warmer
For Use With (Application) Provide safe and nontoxic heat in a format that conforms easily and gently to the infant's heel by using medical tape
Includes Feather edges
Quantity 25 Pack
View More Specs
Products 2
Catalog Number Format Price Quantity & Availability  
Catalog Number Format Price Quantity & Availability  
22-024647 View Documents Gel This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
22-024646 View Documents Liquid This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
The increased blood flow makes fewer skin punctures necessary, which allows for less squeezing, fewer infections, less risk of cartilage damage and faster healing.
  • Provide safe, nontoxic heat in a format that conforms easily and gently to the infant's heel
  • Medical tape allows easy application and removal
  • Easy, reliable flex-disk activation
  • Available in either liquid or gel
  • Consistent 42°C (107°F) temperature provides optimal warming
  • “Feather edges” will not scratch infant's foot
Specifications
Specifications
Flex-disk
Provide safe and nontoxic heat in a format that conforms easily and gently to the infant's heel by using medical tape
25 Pack
42°C
Infant Heel Warmer
Feather edges
107°F