Fisherbrand™ Institutional Trash Can Liners Available on GSA/VA Contract for Federal Government customers only.

Provide a leak resistant containment solution for non-medical waste disposal

 View more versions of this product

Catalog No. 14-828-421

  Check Availability
Add to Cart

Alert

Not for use with biohazardous waste.


    Specifications

    Product Type Institutional Trash Can Liner
    Capacity (English) 56 gal.
    Color Clear
    Guage (English) 12mic
    For Use With (Application) Collecting and containing refuse, non medical waste disposal
    Capacity 56 gal., 211L
    Material HDPE
    Capacity (Metric) 211L
    Thickness 12mic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit