Learn More
Fisherbrand™ Kolmer Serological Pipets Available on GSA/VA Contract for Federal Government customers only.
For Kolmer Complement Fixation technique
 View more versions of this product
Catalog No. 13-676F
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1mL
±0.02mL
To deliver to baseline
0.01mL
Yes
Borosilicate Glass
Yellow
Serological Pipet
Yellow
1/100mL