Learn More
Fisherbrand™ Lab Automation Filter Pipet Tips
Provide maximum performance, accuracy and reproducibility in laboratory automation procedures.
$70.03 - $1128.74
Specifications
Filtered Filtered
Autoclavable Autoclavable
For Use With (Application) Demanding HTS procedures, lab automation procedures; fit several standard and customized automated liquid-handling systems
Format Rack
Tip Style Automation
View More Specs
Products 5
Catalog Number Color For Use With (Equipment) Sterility Volume (Metric) Price Quantity & Availability  
Catalog Number Color For Use With (Equipment) Sterility Volume (Metric) Price Quantity & Availability  
02-683-272
View Documents
Natural Qiagen™ BioRobot™, Qiagen™ Rosys™ Sterile 1100 μL
Pack of 960 for $206.75
Case of 5 PK for $754.43
Add to Cart
02-707-268
View Documents
Black Tecan™ Genesis™ Sterile 50 μL
Pack of 960 for $236.78
Case of 5 PK for $979.65
Add to Cart
02-707-265
View Documents
Natural Tecan™ Genesis™ Sterile 200 μL
Case of 4800 for $1,128.74
Add to Cart
02-707-270
View Documents
Black Tecan™ Genesis™ Sterile 1000 μL
Pack of 960 for $105.63
Case of 5 PK for $528.14
Add to Cart
02-707-241
View Documents
Natural Beckman Coulter™ Biomek™ 1000/2000 Sterile 20 μL
Pack of 960 for $70.03
Case of 5 PK for $350.17
Add to Cart
Description
Description
Tips are constructed of a highly conductive material necessary to detect the presence or absence of sufficient liquid, in order to avoid short sampling and excessive submersion of tip during aspiration.
  • Disposable tips for robotic workstations provide maximum performance, accuracy and reproducibility in laboratory automation procedures
  • Available in clear, blue tint, or carbon-black-conductive polypropylene
  • Carbon-black, liquid-level-sensing tips are required by many automated systems
Specifications
Specifications
Filtered
Demanding HTS procedures, lab automation procedures; fit several standard and customized automated liquid-handling systems
Automation
Polypropylene
Autoclavable
Rack
Yes