Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Fisherbrand™ Lab Automation Large Micron Filter (LMF) Pipet Tips
Provide maximum performance, accuracy and reproducibility in laboratory automation procedures
$190.00
Products 1
Catalog Number Volume (Metric) Price Quantity & Availability  
Catalog Number Volume (Metric) Price Quantity & Availability  
02-707-347
View Documents
150 μL
Pack of 960 for $190.00
Case of 5 PK for $863.06
N/A
Description
Description
For Robotic Workstations: BioTek™ Precision™, Tecan™ Te-Mate. Carbon-black, liquid-level-sensing tips are required by many automated systems. They are constructed of a highly conductive material that is necessary to detect presence or absence of sufficient liquid, in order to avoid short sampling and excessive submersion of tip during aspiration.
  • Ideal for demanding HTS procedures
  • Disposable
  • Engineered to ensure proper fit and function with several standard and customized automated liquid-handling systems
  • Choose tips in an assortment of volume sizes and styles