Learn More
Description
For Robotic Workstations: BioTek™ Precision™, Tecan™ Te-Mate. Carbon-black, liquid-level-sensing tips are required by many automated systems. They are constructed of a highly conductive material that is necessary to detect presence or absence of sufficient liquid, in order to avoid short sampling and excessive submersion of tip during aspiration.
- Ideal for demanding HTS procedures
- Disposable
- Engineered to ensure proper fit and function with several standard and customized automated liquid-handling systems
- Choose tips in an assortment of volume sizes and styles
Specifications
Specifications
| Volume (Metric) | 150 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Natural |
| Autoclavable | Autoclavable |
| For Use With (Application) | Liquid-level-sensing to detect presence or absence of sufficient liquid, provide maximum performance, accuracy and reproducibility in laboratory automation procedures |
| Format | Rack |
| Tip Style | Automation |
| Disposable | Single-use |
| For Use With (Equipment) | BioTek Precision, Tecan TeMo, Tecan GenMate |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
