Learn More
Fisherbrand™ Lab Automation Pipet Tips
Provide maximum performance, accuracy and reproducibility in laboratory automation procedures.
$7.33 - $176.93
Specifications
Filtered Non-Filtered
Autoclavable Autoclavable
Format Rack
Tip Style Automation
Disposable Yes
View More Specs
Products 12
Catalog Number Color Volume (Metric) Sterility For Use With (Equipment) Price Quantity & Availability  
Catalog Number Color Volume (Metric) Sterility For Use With (Equipment) Price Quantity & Availability  
02-707-224
View Documents
Natural 100 μL Non-sterile Zymark™/Caliper™ RapidPlate™ 96, Zymark™/Caliper™ RapidPlate™ 96/384
Pack of 960 for $28.62
Save 67%
Reg : $87.40
The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Learn More
Case of 5 PK for $143.09
Save 59%
Reg : $350.00
The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Learn More
Add to Cart
02-707-246
View Documents
Black 20 μL Non-sterile Tecan™ Genesis™
Pack of 960 for $85.63
Save 64%
Reg : $239.93
The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Learn More
Case of 5 PK for $428.14
Save 61%
Reg : $1,090.96
The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Learn More
Add to Cart
02-707-251
View Documents
Black 50 μL Non-sterile Tecan™ Genesis™
Pack of 960 for $68.81
Case of 5 PK for $388.10
Add to Cart
02-707-254
View Documents
Natural 200 μL Non-sterile Tecan™ Genesis™
Pack of 960 for $108.63
Case of 5 PK for $482.93
Add to Cart
02-707-255
View Documents
Natural 200 μL Sterile Tecan™ Genesis™
Pack of 960 for $7.33
Save 87%
Reg : $54.71
The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Learn More
Case of 5 PK for $36.63
Save 95%
Reg : $730.80
The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Learn More
Add to Cart
02-707-256
View Documents
Black 5 μL to 200 μL Non-sterile Tecan™ Genesis™
Pack of 960 for $168.74
Case of 5 PK for $718.21
Add to Cart
02-707-259
View Documents
Natural 1000 μL Non-sterile Tecan™ Genesis™
Pack of 960 for $164.75
Case of 5 PK for $732.90
Add to Cart
02-707-262
View Documents
Black 1000 μL Sterile Tecan™ Genesis™
Pack of 960 for $105.88
Case of 5 PK for $1,021.12
Add to Cart
02-707-261
View Documents
Black 1000 μL Non-sterile Tecan™ Genesis™
Pack of 960 for $176.93
Case of 5 PK for $743.93
Add to Cart
02-707-219
View Documents
Natural 200 μL Non-sterile Zymark™/Caliper™ RapidPlate™ 96, Zymark™/Caliper™ RapidPlate™ 96/384
Pack of 960 for $99.51
Case of 5 PK for $413.83
Add to Cart
02-707-220
View Documents
Natural 200 μL Sterile Zymark™/Caliper™ RapidPlate™ 96, Zymark™/Caliper™ RapidPlate™ 96/384
Pack of 960 for $120.75
Case of 5 PK for $544.93
Add to Cart
02-707-244
View Documents
Natural 20 μL Non-sterile Tecan™ Genesis™
Pack of 960 for $160.23
Case of 5 PK for $712.42
Add to Cart
Description
Description
Tips are constructed of a highly conductive material necessary to detect the presence or absence of sufficient liquid, in order to avoid short sampling and excessive submersion of tip during aspiration.
  • Disposable tips for robotic workstations provide maximum performance, accuracy and reproducibility in laboratory automation procedures
  • Available in clear, blue tint, or carbon-black-conductive polypropylene
  • Carbon-black, liquid-level-sensing tips are required by many automated systems
Specifications
Specifications
Non-Filtered
Rack
Yes
Autoclavable
Automation
Polypropylene