Learn More
Fisherbrand™ Lab Automation Nonfilter Pipet Tips
Provide maximum performance, accuracy and reproducibility in laboratory automation procedures
 View more versions of this product
Catalog No. 02-707-246
Add to cart
Description
Description
Carbon-black, liquid-level-sensing tips are required by many automated systems. They are constructed of a highly conductive material that is necessary to detect presence or absence of sufficient liquid, in order to avoid short sampling and excessive submersion of tip during aspiration.
Specifications
Specifications
Tecan™ Genesis™
Automation
Non-Filtered
Black
PP (Polypropylene)
Liquid-level-sensing to detect presence or absence of sufficient liquid, provide maximum performance, accuracy and reproducibility in laboratory automation procedures
Rack
20μL
Non-sterile
Yes
Yes
Tecan™ Genesis™