Fisherbrand™ Lab Automation Nonfilter Pipet Tips Available on GSA/VA Contract for Federal Government customers only.

Provide maximum performance, accuracy and reproducibility in laboratory automation procedures

 View more versions of this product

Catalog No. 02-707-249

  Check Availability
Add to Cart

    Specifications

    Product Type Robotic Tip
    Material Polypropylene
    Quantity 96 Tips/Rack
    For Use With (Equipment) Tecan™ Genesis™ robotic lab automation workstation
    Sterility Nonsterile
    Autoclavable Yes
    Packaging Rack
    For Use With (Application) Provide maximum performance, accuracy, and reproducibility in laboratory automation procedures
    Volume (Metric) 50μL
    Color Natural

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit