Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Lab Automation Nonfilter Pipet Tips-Zymark RapidPlate 96and 96/384-Bulk
GREENER_CHOICE

Catalog No. 02707218 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
200 μL

Provide maximum performance, accuracy, and reproducibility in laboratory automation procedures

  • Volume: 200μL
  • Non-sterile
  • Non-filtered
  • Color: Natural
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
For Use With (Application) Provide maximum performance, accuracy, and reproducibility in laboratory automation procedures
Format Rack
Tip Style Automation
Disposable Single-use
For Use With (Equipment) Zymark/Caliper RapidPlate 96, Zymark/Caliper RapidPlate 96/384
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.