Learn More
Fisherbrand™ Lab Automation Nonfilter Pipet Tips
Provide maximum performance, accuracy and reproducibility in laboratory automation procedures
 View more versions of this product
Catalog No. 02-707-225
Add to cart
Description
Description
Carbon-black, liquid-level-sensing tips are required by many automated systems. They are constructed of a highly conductive material that is necessary to detect presence or absence of sufficient liquid, in order to avoid short sampling and excessive submersion of tip during aspiration.
Specifications
Specifications
Zymark™/Caliper™ RapidPlate™, Zymark™/Caliper™ RapidPlate™
Automation
Non-Filtered
Natural
PP (Polypropylene)
Provide maximum performance, accuracy, and reproducibility in laboratory automation procedures
Rack
100μL
Sterile
Yes
Yes
Zymark™/Caliper™ RapidPlate™ 96, Zymark™/Caliper™ RapidPlate™ 96/384