Learn More
Fisherbrand™ Lab-Jacks™
Nonsparking anodized aluminum plates for maximum safety in the lab
 View more versions of this product
Catalog No. 14-673-51
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
59 lb.
2.45 to 11 in.
6 in.
Stainless Steel, Anodized Aluminum Top
6 in.
15cm
Supporting hotplates, baths, glassware
6.2 to 27.3cm
15cm
Lab Jack
6 in.
15cm