Fisherbrand™ Lighted Refractometers
Integrated LED ensures that your samples are always clearly visible
 View more versions of this product
Catalog No. 12-561-322
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Accuracy 0.2°
    Scale Range 0 to 32° Brix
    For Use With (Application) Sugar related liquids in juice, wine, soft drinks, other beverages, honey and food
    Includes Nylon padded zipper pouch, pipette and screwdriver
    Lighting Type Integrated LED
    Temperature Compensation Automatic
    Weight (Metric) 176 grams
    The “L” series offers premium features, build quality, and durability at a cost only marginally higher than our industrial and economy models.
    • Integrated LED light source
    • Rugged, rubber-coated outer construction
    • Self-stowing sample sticks and lanyard
    • Ruggedized, drop-tested daylight plate
    • Magnified scale for precise readings
  • Description & Specifications
Related Products
Product Content Feedback