Fisherbrand™ Lighted Refractometers
Integrated LED ensures that your samples are always clearly visible
 View more versions of this product
Catalog No. 12-561-321
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Includes Nylon padded zipper pouch, pipette and screwdriver
    Lighting Type Integrated LED
    Temperature Compensation Automatic
    Weight (Metric) 176 grams
    Accuracy 0.1°
    Scale Range 0 to 10° Brix
    For Use With (Application) Sugar related liquids in juice, wine, soft drinks, other beverages, honey and food
  • Description & Specifications
Related Products
Product Content Feedback