Fisherbrand™ Locking Dialysis Membrane Clamps
 View more versions of this product
Catalog No. 21-153-100
  Check Availability
Add to Cart
    Specifications
    Quantity 10 Pack
    For Use With (Application) Do not float, stay locked until released
    Material Polyamide 6.6 Dupont (Nylon)
    Product Type Dialysis Membrane Clamp, Up to 46mm sealing width
  • Description & Specifications
Related Products
Product Content Feedback