Fisherbrand™ Locking-Lid Microcentrifuge Tubes with Polypropylene Snap-Cap™ Available on GSA/VA Contract for Federal Government customers only.
Convenient, pierceable locking lids snap into place with one-handed operation
 View more versions of this product
Catalog No. 02-681-277
  Check Availability
Add to Cart
    Specifications
    Capacity (Metric) 0.6mL
    Color Yellow
    Material Polypropylene
    Max. Force 30000xG
    For Use With (Application) Ideal for sample-handling applications, including centrifugation, boiling and freezing of specimens, incubation and storage
    Graduations 0.1mL
    Packaging Bag
    Certifications/Compliance RNase, DNase-free
    Closure Material Polypropylene
    Closure Type Snap
    Autoclavable Yes
  • Description & Specifications
Related Products
Product Content Feedback