Learn More
Fisherbrand™ Low-Form Cylinders with Double Metric Scales, Capacity Available on GSA/VA Contract for Federal Government customers only.
Calibrated “To Deliver”
 View more versions of this product
Catalog No. 08-562B
Add to cart
Description
Description
Calibrated “To Deliver”
  • Two white scales for reading up or down
  • Accuracy assured even at lowest graduations
  • Won't easily tip
  • Two pouring lips
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
150mm
100mL
5.9 in.
±10mL
Yes
2mL
Borosilicate Glass
Accuracy assured even at lowest graduations
Double metric scale
2mL
+/- 1.5mL