Learn More
Fisherbrand™ Low-Retention Pipet Tips - Filtered
Increase accuracy by reducing sample retention
 View more versions of this product
Catalog No. 02-717-160
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
  • Low retention surface minimizes samples loss
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
  • Natural color
Description & Specifications
Specifications
20μL
Natural
Filter
Eppendorf Pipetters
Sterile
Low retention
Racked: 10 x 100